Silver, brazing alloy, Ø 0.8 mm (0.03 in)
Catalog # 41026.15
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:30 g
- Seq:3332
Specifications
About this item
:
Storage Temperature:Ambient
Formula:Ag:Cu:Zn:Sn;56:22:17:5%
MDL Number:MFCD00801120