Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dicarbonylbis(cyclopentadienyl)titanium(II)
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
39571.02
:
Thermo Fisher Scientific
:
Dicarbonylbis(cyclopentadienyl)titanium(II) ≥98%
39571.02
:
Thermo Fisher Scientific
:
Pk:
0,5 g
Seq:
3035
:
:
234.076 g/mol
:
60 °C (0,001 mmHg) (subl.)
:
90 °C (dec.)
:
6.1,II
:
GHS02
:
GHS06
:
Ambient
:
12129-51-0
:
C₁₂H₁₀O₂Ti
:
MFCD00143762