

Nickel(II) octanoate (8% Ni) in mineral spirits
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4995-91-9
:
Formula:C₁₆H₃₀NiO₄
MDL Number:MFCD00064239
:345.10 g/mol
Density:0,95
Flash Pt:40 °C (104 °F)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS08
:GHS02
Related Information
Frequently Bought Together