Cerium zirconium oxide ≥99.5% (metals basis)
Catalog # 39216.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
:
:295.32 g/mol
Storage Temperature:Ambient
EINECS:258-407-6
CAS Number:53169-24-7
Formula:CeO₂·ZrO₂
MDL Number:MFCD00168089