£0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Northern Ireland
ContactUsLinkComponent
Lanthanum (III) chloride
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
39036.AP
:
Thermo Fisher Scientific
:
Lanthanum(III) chloride 1% La (La2O3) in HCl, Specpure®
39036.AP
:
Thermo Fisher Scientific
:
Pk:
500 mL
Seq:
2725
:
:
245.26 g/mol
:
Ambient
:
231-595-7
:
3264
:
8,III
:
GHS05
:
10099-58-8
:
Cl₃La
:
MFCD00011068