
Brucine sulphate heptahydrate ACS
Catalog # 36249.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:887.01 g/mol
Merck Index:14,1455
Melting Pt:180 °C (dec.)
Storage Temperature:Ambient
EINECS:225-432-9
UN:2811
ADR:6.1,I
:GHS06
CAS Number:60583-39-3
Formula:C₂₃H₂₆N₂O₄
MDL Number:MFCD00013472