
N,N-Bis(2-hydroxyethyl)ethylenediamine
Catalog # 32312.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:148.206 g/mol
Boiling Pt:174…178 °C (3…4 mmHg)
Density:1,069
ADR:8,III
:GHS05
Storage Temperature:Ambient
CAS Number:3197-06-6
Formula:C₆H₁₆N₂O₂
MDL Number:MFCD00014822