Iron(II) ethylenediammonium sulphate tetrahydrate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Ferrous Ethylenediamine Sulphate
CAS Number:113193-60-5
:34962-29-3
Formula:C₂H₈N₂
MDL Number:MFCD00082474
:310.07 g/mol
Storage Temperature:Ambient
EINECS:264-357-6
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together