
Ammonium tetrathiotungstate
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13862-78-7
:
Formula:H₈N₂S₄W
MDL Number:MFCD00061410
:348.158 g/mol
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together