
Ammonium molybdate (para) hydrate ≥99.999% (metals basis excluding W), Puratronic® (max. 300 ppm W)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12027-67-7
:
Formula:H₂₄Mo₇N₆O₂₄·xH₂O
MDL Number:MFCD00064633
:1163.8 g/mol (anhy)
Merck Index:14,533
Melting Pt:∼ 190 °C (dec.)
Density:2,498
EINECS:234-722-4
:GHS07
Storage Temperature:Ambient
Frequently Bought Together