
Rubidium formate hydrate ≥99.8% (metals basis)
Catalog # 13499.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:130.49 g/mol (anhy)
:GHS07
Storage Temperature:Ambient
CAS Number:3495-35-0
Formula:CHO₂Rb·xH₂O
MDL Number:MFCD00054359