
Dichloro(1,5-cyclooctadiene)platinum(II) (51,6 - 52,6% Pt)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:12080-32-9
:
Formula:C₈H₁₂Cl₂Pt
MDL Number:MFCD00012413
:374.17 g/mol
Melting Pt:250 °C (dec.)
EINECS:235-144-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together