L(+)-Ascorbic acid ≥99.0%, AR® ACS, Macron Fine Chemicals™
Catalog # 4407-04
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle, amber, wide mouth
- Pk:500 g
- Seq:3645
- Environmentally Preferable:
Specifications
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≥99.0%
- For Laboratory,Research,or Manufacturing Use:Passes Test
- ACS - Assay (C₆H₈O₆):≥99.0 %
- ACS - Appearance (colorless or white to slightly yellow crystals):Passes Test
- Heavy Metals (as Pb):≤0.002 %
- Trace Impurities - Iron (Fe):≤0.001 %
- Residue on Ignition:≤0.10 %
- Specific Rotation, [Å]²⁵^D(c=10 in water)(+):20.5 - 21.5 Degree
Specification Test Results
About this item
Synonyms: Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
:
:176.13 g/mol
Boiling Pt:553
Melting Pt:190…192 °C
Density:1.694 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-066-2
REACH:exempt
CAS Number:50-81-7
Formula:C₆H₈O₆
MDL Number:MFCD00064328
Merck Index:13,00837