Order Entry
Northern Ireland
ContactUsLinkComponent
n-Hexadecane, J.T.Baker®
Health
n-Hexadecane, J.T.Baker®
Catalog # N105-07
Supplier:  Avantor
CAS Number:  
undefined
Health
n-Hexadecane, J.T.Baker®
Catalog # N105-07
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Identification (by IR):
    Passes Test

Specification Test Results

About this item

 :
 :226.45 g/mol
Boiling Pt:287 °C (1013 hPa)
Melting Pt:18 °C
Density:0,77331 g/cm³ (20 °C)
Flash Pt:135 °C (closed cup)
Storage Temperature:Ambient
EINECS:208-878-9
 :GHS08
CAS Number:544-76-3
Formula:H₃C(CH₂)₁₄CH₃
MDL Number:MFCD00008998