About this item
The Biosensis Neurotrophin 4/5 Rapid™ enzyme-linked immunosorbent assay (ELISA) kit is a sandwich ELISA that allows the specific, fast and reliable quantification of NT4/5 in less than 4 hours in cell culture supernatants, human citrate-plasma and brain extracts only if used as directed.
This NT4/5 ELISA kit employs a recombinant human NT4/5 standard. The purpose of this kit is the in vitro quantitative determination of human NT4/5 in samples such as cell culture supernatants, serum and plasma (EDTA and citrate). The capture and detection antibodies will also detect NT4/5 from other species due to a high degree of NT4/5 amino acid sequence homology. Therefore, this ELISA kit can be used to quantify NT4/5 in many species including mouse, rat and monkey.
The antibodies used in this kit bind to epitopes within the mature domain of NT4/5. Thus, this ELISA kit will quantify the mature form of NT4/5, and may also detect the full-length pro-form of NT4/5.
This kit consists of a pre-coated polyclonal anti-NT4/5 capture antibody, a biotinylated anti-NT4/5 detection antibody and horseradish peroxidase (HRP)-conjugated streptavidin. The addition of a substrate (3,3’,5,5’-tetramethylbenzidine, TMB) yields a coloured reaction product, which is directly proportional to the concentration of NT4/5 present in samples and protein standards.
Please refer to the kit protocol for specific use instructions for each substrate application, in particular blood samples.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Assay Type:Sandwich
- Primary antibody reactivity:HumanRatMouse
- Detection range:15,6 - 1000 pg/ml
- Sensitivity:Typically detects down to 10 pg/ml human NT4/5 (defined as 150% of blank value)