Order Entry
Northern Ireland
ContactUsLinkComponent
Fluorescent indicator F₂₅₄, Millipore®
Fluorescent indicator F₂₅₄, Millipore®
Catalog # 1.09182.0050
Supplier:  Merck
CAS Number:  
undefined
Fluorescent indicator F₂₅₄, Millipore®
Catalog # 1.09182.0050
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    50 g
  • Seq:
    3368

Specifications

Specification Test Results

  • Identity:
    Conforms
  • Fe (iron):
    ≤0,01%
  • Water:
    ≤0,3%
  • Application test for DC:
    Conforms

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:271-896-0
CAS Number:68611-47-2