Aminoacetonitrile sulphate for synthesis, Sigma-Aldrich®
8.41034.0050
: Merck
:
- Pack type:Glass bottle
- Pk:50 g
- Seq:3368
- Melting Point:118 - 122 °C
- Molar Mass:210.2 g/mol
- Synonyms:Glycinonitrile sulphate
- Hill Formula:C4H10N4O4S
- Storage temperature:+15...+25 °C
:
:210.21 g/mol
:6.1,III
:M-Clarity™ quality level = MQ200
:Ambient
:226-772-0
:3439
:5466-22-8
:MFCD01941514
:C₄H₁₀N₄O₄S
:MFCD00012851