Order Entry
Northern Ireland
ContactUsLinkComponent
 
Celite® 281 pH 10 (100 g/l, H₂O, 20 °C), Supelco®
Catalog # 1.02686.1000
Supplier:  Merck
CAS Number:  
Celite® 281 pH 10 (100 g/l, H₂O, 20 °C), Supelco®
Catalog # 1.02686.1000
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Solubility:
     - 0.001 g/l (20 °C)
  • Bulk Density:
    300 kg/m3
  • pH:
    10 (100 g/l, H2O, 20 °C)
  • pH value:
    10
  • Identity:
    Passes test
  • Loss on ignition:
    ≤2,0 %

Specification Test Results

About this item

 :
 :M-Clarity™ quality level = MQ100
EINECS:272-489-0
 :GHS08
Storage Temperature:Ambient
CAS Number:68855-54-9