(+)-Ethyl-4,6-di-O-acetyl-2,3-dideoxy-α-D-erythro-hex-2-enopyranoside for synthesis
Catalog # 8.18289.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:76 - 78 °C
- Molar Mass:258.27 g/mol
- Bulk Density:200 kg/m3
- Hill Formula:C12H18O6
- Storage:+15...+25 °C
- Assay (DSC):≥99,0%
- Specific rotation:105,0 - 107,0 °
- Identity (IR):Passes test
Specification Test Results
About this item
:
:258.27 g/mol
:M-Clarity™ quality level = MQ200
CAS Number:3323-72-6
Formula:C₁₂H₁₈O₆
MDL Number:MFCD01103303