Order Entry
Northern Ireland
ContactUsLinkComponent
2-Naphthyl phenyl ketone for synthesis, Sigma-Aldrich®
2-Naphthyl phenyl ketone for synthesis, Sigma-Aldrich®
Catalog # 8.20869.0001
Supplier:  Merck
CAS Number:  
2-Naphthyl phenyl ketone for synthesis, Sigma-Aldrich®
Catalog # 8.20869.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

Specification Test Results

  • Solubility:
    (20 °C) insoluble
  • Melting Point:
    80 - 82 °C
  • Molar Mass:
    232.28 g/mol
  • Boiling Point:
    398 °C
  • Synonyms:
    Phenyl 2-naphthyl ketone
  • Hill Formula:
    C17H12O

Specification Test Results

About this item

 :
 :232.28 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:211-410-6
CAS Number:644-13-3
Formula:C₁₇H₁₂O
MDL Number:MFCD00004106