Order Entry
Northern Ireland
ContactUsLinkComponent
Formamidine disulphide dihydrochloride for synthesis, Sigma-Aldrich®
Formamidine disulphide dihydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.14609.0010
Supplier:  Merck
CAS Number:  
undefined
Formamidine disulphide dihydrochloride for synthesis, Sigma-Aldrich®
Catalog # 8.14609.0010
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 g
  • Seq:
    3236

Specifications

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    175 - 179 °C
  • Molar Mass:
    223.15 g/mol
  • Storage:
    +2...+8 °C
  • Assay (argentometric):
    ≥97,0% (m)
  • Identity:
    Conforms

Specification Test Results

About this item

 :
 :223.13 g/mol
 :M-Clarity™ quality level = MQ200
Storage Temperature:Refrigerator
EINECS:238-876-3
CAS Number:14807-75-1
Formula:C₂H₆N₄S₂
MDL Number:MFCD00012586