Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Vpr (HIV-1) Binding Protein
- Antigen symbol:VPRBP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:9730
- Antigen synonyms:DCAF1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
P55 is an extensively palmitoylated erythrocyte membrane protein, and a member of the MAGUK family. P55 also resists salt extraction, resulting in a high affinity for the plasma membrane. P55 contains a PDZ/DHR domain, a conserved SH-3 domain that appears to suppress tyrosine kinase activity of various oncoproteins, a 39-amino acid motif that binds to cytoskeletal protein 4.1R, and a guanylate kinase-like domain. Interaction with glycophorin C (GPC) and 4.1R suggests that p55 may play a role in the dynamic regulation in the erythrocyte membrane. In addition, p55 gene expression in vivo may be associated with a CpG island. P55 is constitutively expressed in K562 erythroleukemia cells during erythropoiesis and undergoes a 2-fold amplification after induction.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: VPRBP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat