Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-SLFN14 Rabbit Polyclonal Antibody
Anti-SLFN14 Rabbit Polyclonal Antibody
  BOSSBS-9190R
 :  Bioss
Anti-SLFN14 Rabbit Polyclonal Antibody
  BOSSBS-9190R
 :  Bioss

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Schlafen Family Member 14
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    liquid
  • Gene ID:
    342618
  • Antigen synonyms:
    Schlafen family member 14|SLFN14|SLN14_HUMAN.
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Concentration:
    1 ug/ul
  • Shipping temperature:
    4 °C
  • Purification:
    Purified by Protein A.
  • Size:
    100 μg
  • Pk:
    100 µl

 

 

Schlafen family members are preferentially expressed in lymphoid tissues and are differentially regulated during thymocyte maturation. Schlafen proteins function as suppressors of cell growth and are thought to play a role in the maintenance of T cell quiescence. The prototype member of the Schlafen family, Slfn1, is transcriptionally unregulated during thymocyte positive selection, and the induction of Slfn1 induces a G0/G1 arrest, suggesting that Slfn1 participates in the regulation of cell cycle and potentially acts as a determining factor for apoptosis. These proteins all contain a largely conserved core domain within the center of the sequence, and yet they are substantially diversified at the N terminus. Slfn14 (Schlafen family member 14) is a 912 amino acid protein belonging to the Schlafen family. Slfn14 exists as two alternatively spliced isoforms and is encoded by a gene mapping to human chromosome 17q12.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: SLFN14
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat