Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:DNA methyltransferase 1 associated protein 1
- Antigen symbol:DMAP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Gene ID:55929
- Antigen synonyms:DNMAP1|FLJ11543|DKFZp686L09142|SWC4|MEAF2|DNMTAP1|KIAA1425|EAF2
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
- Concentration:1 ug/ul
- Shipping temperature:4 °C
- Purification:Purified by Protein A.
- Size:100 μg
- Pk:100 µl
Specifications
About this item
Methylation of DNA contributes to the regulation of gene transcription in eukaryotic systems. DNA methylation is predominantly found on cytosine residues that are present in dinucleotide motifs consisting of a 5' cytosine followed by a guanosine (CpG), and it requires the enzymatic activity of DNA methyltransferases (DNMTs), which results in transcriptional repression of the methylated gene. DNA methyltransferase 1-associating protein (Dmap1) binds to methyl-CpG rich domains and mediate the transcriptional inhibition associated with DNA methylation. Dmap1 interacts with Daxx to enhanced Daxx-mediated repression of glucocorticoid receptor transcriptional activity. Daxx also protects Dmap1 from protein degradation in vivo.
WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)
Type: Primary
Antigen: DMAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat