(-)-Menthyl acetate for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2623-23-6
:
Formula:C₁₂H₂₂O₂
MDL Number:MFCD00001482
:198.31 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:220-076-0
Specification Test Results
- Molar Mass:198.3 g/mol
- Boiling Point:229.5 °C
- Density:0.925 g/cm3
- Hill Formula:C12H22O2
- Assay (GC, area%):≥97 Area%
- Identity (IR):Passes test
- Storage temperature:+15...+25 °C
Specification Test Results
Frequently Bought Together