Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-SERPINI1 Rabbit Polyclonal Antibody
  BSBTPB9744
undefined
Anti-SERPINI1 Rabbit Polyclonal Antibody
  BSBTPB9744

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    Serpin peptidase inhibitor, clade I, member 1
  • Antigen symbol:
    SERPINI1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    5274
  • Antigen synonyms:
    PI12|neuroserpin
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Neuroserpin(SERPINI1) detection. Tested with WB in Human;Mouse;Rat.

Type: Primary
Antigen: SERPINI1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat