Order Entry
Northern Ireland
ContactUsLinkComponent
 
Isopropyl 2-bromo-2-methylpropionate for synthesis, Sigma-Aldrich®
 :  Merck
 
Isopropyl 2-bromo-2-methylpropionate for synthesis, Sigma-Aldrich®
 :  Merck

 

  • Solubility:
    (20 °C) insoluble
  • Molar Mass:
    209.08 g/mol
  • Boiling Point:
    170 °C
  • Flash Point:
    63 °C
  • Density:
    1.24 g/cm3 (20 °C)
  • Assay (GC, area%):
    ≥ 98% (a/a)
  • Storage temperature:
    +15...+25 °C
  • Identity (IR):
    Passes test

 

 

Frequently Bought Together