Rotary evaporator splash head
Catalog # SMIT8SH1524
Supplier: SMITH SCIENTIFIC
Specifications
- Cone size:24 NS
- Capacity:100 ml
- Pk:100 mL
Specifications
About this item
Borosilicate glass 3.3
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes