Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-6x His Mouse Monoclonal Antibody (DyLight® 650) [clone: AD1.1.10]
  AGRIAS152933
 :  Agrisera
Anti-6x His Mouse Monoclonal Antibody (DyLight® 650) [clone: AD1.1.10]
  AGRIAS152933
 :  Agrisera

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antibody type:
    Primary
  • Antigen name:
    His Tag
  • Antigen symbol:
    6xHis
  • Clonality:
    Monoclonal
  • Clone:
    AD1.1.10
  • Conjugation:
    DyLight® 650
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Reactivity:
    Birds,
    Bovine
  • Immunogen:
    Polyhistidine (HHHHHH) tagged polypeptide
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight®650. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.,Mouse anti-6x Histidine is a primary antibody which binds to 6x Histidine tag and is directly conjugated to DyLight®650. This is of advantage to shorten assay time by no need to use a secondary antibody to 6x Histidine tag. DyLight® is a registered trade mark of Thermofisher Inc., and its subsidaries.

Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: AD1.1.10
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype:
Reactivity: Birds, Bovine