Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-MAPK6 Rabbit Polyclonal Antibody
  BSBTPB9666
undefined
Anti-MAPK6 Rabbit Polyclonal Antibody
  BSBTPB9666

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antigen name:
    Mitogen-activated protein kinase 6
  • Antigen symbol:
    MAPK6
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Gene ID:
    5597
  • Antigen synonyms:
    PRKM6|ERK3|HsT17250|p97MAPK
  • UniProtKB:
    Q16659
  • Storage temperature:
    At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
  • Immunogen:
    E.coli-derived human MAPK6 recombinant protein (Position: E520-N721). Human MAPK6 shares 86.2% and 82.8% amino acid (aa) sequence identity with mouse and rat MAPK6, respectively.
  • Purification:
    Immunogen affinity purified.
  • Size:
    100 μg / vial
  • Pk:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Mitogen-activated protein kinase 6(MAPK6) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Mitogen-activated protein kinase 6 is an enzyme that in humans is encoded by the MAPK6 gene. It is mapped to 15q21. The protein encoded by this gene is a member of the Ser/Thr protein kinase family, and is most closely related to mitogen-activated protein kinases (MAP kinases). MAP kinases also known as extracellular signal-regulated kinases (ERKs), are activated through protein phosphorylation cascades and act as integration points for multiple biochemical signals. This kinase is localized in the nucleus, and has been reported to be activated in fibroblastsupon treatment with serum or phorbol esters.

Add 0.2ml of distilled water will yield a concentration of 500ug/ml.

Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.

Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.