Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CTGF Rabbit Polyclonal Antibody
Anti-CTGF Rabbit Polyclonal Antibody
Catalog # BSBTPB9541
Supplier:  Boster Bio
undefined
Anti-CTGF Rabbit Polyclonal Antibody
Catalog # BSBTPB9541
Supplier:  Boster Bio

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    connective tissue growth factor
  • Antigen symbol:
    CTGF
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Gene ID:
    1490
  • Antigen synonyms:
    IGFBP8|HCS24|CCN2|NOV2
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Connective tissue growth factor(CTGF) detection. Tested with WB, IHC-P in Human;Rat.

Type: Primary
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat