Order Entry
Northern Ireland
ContactUsLinkComponent
 
Anti-HLA-DPB1 Mouse Monoclonal Antibody
  ABNOH00003115-PW1
 :  Abnova
Anti-HLA-DPB1 Mouse Monoclonal Antibody
  ABNOH00003115-PW1
 :  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Antigen name:
    HLA class II histocompatibility antigen, DP(W2) beta chain
  • Antigen symbol:
    HLA-DPB1
  • Clonality:
    Monoclonal
  • Host:
    Mouse
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    3115
  • Antigen synonyms:
    DPB1|HLA-DP|HLA-DPB|HLA-DP1B
  • Storage temperature:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Size:
    1 Set
  • Pk:
    1 SET

 

 

This Immunopreciptitation/ Western Blot Antibody Pair set comes with one Antibody for Immunoprecipitation and another to detect the precipitated protein in Western Blot.

Antibody pair set content: 1. Antibody pair for IP: Mouse Monoclonal Anti-HLA-DPB1 (300 ?g); 2. Antibody pair for Western Blot: Mouse Monoclonal Anti-HLA-DPB1 (50 ?g)

Type:
Antigen: HLA-DPB1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human