Order Entry
Northern Ireland
ContactUsLinkComponent
Analogue stopwatch, Mega Minute
Analogue stopwatch, Mega Minute
  609-0148
 :  HANHART
undefined
Analogue stopwatch, Mega Minute
  609-0148
 :  HANHART

 

  • Ø:
    55 mm
  • Colour:
    Chrome
  • Description:
    Mega Minute, lever, 7 jewels, 1/5 s, 60 min, 125 g
  • Environmentally Preferable:
  • Pk:
    1

 

 

Stopwatch with sprung central minutes hand for measuring up to 60 minutes.

  • Chrome plated and polished brass casing, diamond cut
  • 1/5 second recorder
  • Excellent readability thanks to double-sized minute scale in contrasting colour, and sprung minute hand
  • Precision lever movements (7 jewels)
  • Shock protection, additional dust and water protection