Photometer, MD 610
705-0923
: Lovibond
- Description:MD 610, portabe photometer
- Model:MD 610
- Wavelength accuracy:± 1 nm
- Source lamp:LED
- Display:LCD, backlit
- Filter type:Interference
- Interfaces:Bluetooth
- Optics:LED, interference filters (IF) and photo sensor in transparent sample chamber
- Photometric accuracy:2% FS (T = 20...25°C)
- Power supply:4× mignon AA/LR6 batteries
- Storage capacity:500 data sets
- Wavelengths:430, 530, 56 0, 580, 610, 660 nm
- Weight:~0,45 kg
- W×D×H:210×95×45 mm
- Pk:1
The MD 610 combines the mobility of a handheld photometer with the characteristics of a modern laboratory photometer. This instrument has over 120 pre-programmed methods covering the important methods for water analysis, including COD, chlorine, nutrients and metals. The high level of accuracy of Lovibond® reagents and the user friendly nature of the instrument guarantee rapid and reliable analysis of your water samples. Depending on the application, the unit will operate with tablet reagents, powder packs, liquid reagents or tube tests (¹⁶/₁₃ mm).
- Bluetooth® data transmittance to smartphones and tablets
- Advanced data management via free AquaLX® App for Android and iOS
- High quality results due to interference filters and long-life LEDs
- Automatic wavelength selection
- 35 user defined methods
- Waterproof housing
- User interface in German, English, French, Spanish, Italian, Portuguese (BR), Polish and Indonesian
: Supplied with carrying case, 4×AA/LR6 batteries, 3× round glass vials 13 mm, 3× round glass vials 16 mm,1 adapter each for 16 mm and 13 mm vials, plastic stirring rod, brush and screw driver.