ContactUsLinkComponent
Wavelength calibration solution, concentrate, for ICP-OES and MP-AES
Wavelength calibration solution, concentrate, for ICP-OES and MP-AES
Catalog # VARI6610030000
Supplier:  VARIAN
CAS Number:  
Wavelength calibration solution, concentrate, for ICP-OES and MP-AES
Catalog # VARI6610030000
Supplier:  VARIAN
CAS Number:  
Specifications
  • Pk:
    500 mL
  • Description:
    Wavelength calibration solution, concentrate, for ICP-OES and MP-AES
Specifications
About this item
500 ml solution contains 50 mg/L Al, As, Ba,Cd, Co, Cr, Cu, Mn, Mo, Ni, Pb, Se, Sr, Zn and 500 mg/L K in 5% HNO₃ + tr HF. Solution must be diluted 10 times prior to use.
  • Concentrate for dilution
  • Packaged in pre-leached HDPE bottles, secured with a tamper-evident seal