Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Chromatography Columns
Guard columns, ChromSep®
Guard columns, ChromSep®
Guard columns, ChromSep®
Catalog # VARICP28151
Supplier:
VARIAN
Expand image
Guard columns, ChromSep®
Catalog # VARICP28151
Supplier:
VARIAN
Specifications
Column Format:
Guard column
Ø int.:
4,60 mm
Pore size:
120 Å
Length:
50 mm
Pk:
5
Specifications
About this item
Product Details
Product Details