Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-Neurotrophin-4/5; NT4/NT5 Chicken Polyclonal Antibody
Anti-Neurotrophin-4/5; NT4/NT5 Chicken Polyclonal Antibody
Catalog # BSENC-1514-500
Supplier:  Biosensis
undefined
Anti-Neurotrophin-4/5; NT4/NT5 Chicken Polyclonal Antibody
Catalog # BSENC-1514-500
Supplier:  Biosensis

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Neurotrophin 4/5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Chicken
  • Isotype:
    IgY
  • Reactivity:
    Human,
    Mouse
  • Epitope:
    Human NT4/5 peptides, amino acids 104-118 and 198-210
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Antigen synonyms:
    NTF5|NTF4,
  • Immunogen:
    Mixture of two human NT4/NT5 peptides (104-118 and 198-210 aa).
  • Size:
    500 µl
  • Pk:
    500 µl

Specifications

About this item

NT4 is a member of the neurotrophin family, in knockout animals NT4 deficient mice appeared little changed from wildtype animals, however NT4 is a powerful neurotrophic factor for periferial neuronal ganglia and is capable if sustaining ganglia neurons in vitro. NT4 is closely related to both NGF and BDNF. FUNCTION: Target-derived survival factor for peripheral sensory sympathetic neurons. SUBCELLULAR LOCATION: Secreted protein. TISSUE SPECIFICITY: Highest levels in prostate, lower levels in thymus, placenta, and skeletal muscle. Expressed in embryonic and adult tissues. SIMILARITY: Belongs to the NGF-beta family.

The antibody reacts with human and mouse NT-4//5 proteins and likely with other species based upon homology. The antibody has not been tested in IHC or western blot
Application Information:
ELISA at suggested dilution of 1:500-1:2,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: Neurotrophin-4/5; NT4/NT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human NT4/5 peptides, amino acids 104-118 and 198-210
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse