Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:elF2c Protein
- Antigen symbol:ELF2C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-terminus of Mouse elF2C
- Cross adsorption:No
- Format:Lyophilized
- Antigen synonyms:Ago1|mAgo2|eIF2C 2|Eukaryotic translation initiation factor 2C|eIF-2C 2|Protein argonaute-1|Ago2|Eif2c2|eIF-2C 1
- Immunogen:A synthetic peptide corresponding to a sequence (845-860 aa) from the C-terminus of mouse elF2C conjugated to KLH.
- Size:100 µl
- Pk:100 µl
Specifications
About this item
The elF2c (Argonaute, Ago) protein is a core protein of the RNA-induced silencing complex (RISC).
Specificity has been confirmed by IHC, WB and IP in mouse brain. This antibody is expected to recognise at least elF2c-1, 2 and 3 since they have identical C-termini.
Application Information:
Western Blotting (WB), Immunochemistry (IHC) and Immunoprecipitation (IP). The recommended concentration for IHC in formalin fixed and paraffin embedded tissues and formalin/acetone fixed tissues is 1:200-1:500. For WB, the recommended concentration is 1:1,000 to 1:2,000. The elFC2 protein has a predicted length of 860 residues and the MW of the monomer is 94 kDa. Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: mouse elF2c Protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminus of Mouse elF2C
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat