Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-S100 A8 Mouse Polyclonal Antibody
Anti-S100 A8 Mouse Polyclonal Antibody
Catalog # ABNOH00006279-B02P
Supplier:  Abnova
Anti-S100 A8 Mouse Polyclonal Antibody
Catalog # ABNOH00006279-B02P
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100 calcium binding protein A8
  • Antigen symbol:
    S100A8
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Liquid
  • Gene ID:
    6279
  • Antigen synonyms:
    L1Ag|CP-10|CFAG|CAGA|MRP8|NIF|MIF|CGLA|60B8AG|MA387|P8
  • Amino acid number:
    1 to 93
  • Storage buffer:
    In 1x PBS, pH 7,4
  • Sequence:
    MLTELEKALNSIIDVYHKYSLIKGNFHAVYRDDLKKLLETECPQYIRKKGADVWFKELDINTDGAVNFQEFLILVIKMGVAAHKKSHEESHKE
  • Storage temperature:
    Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    S100A8 (NP_002955.2, 1 a.a. ~ 93 a.a) full-length human protein.
  • Size:
    50 μg
  • Pk:
    50 µg

Specifications

About this item

Mouse polyclonal antibody raised against a full-length human S100A8 protein.

Recommended Dilutions: Western Blot: 1 ?g/ml

Type: Primary
Antigen: S100 A8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: