Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-CACNA1G Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S178A-9]
Anti-CACNA1G Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S178A-9]
Catalog # ABNOMAB11560
Supplier:  Abnova
Anti-CACNA1G Mouse Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: S178A-9]
Catalog # ABNOMAB11560
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Calcium Channel, Voltage-dependent, T type, alpha 1G subunit
  • Antigen symbol:
    CACNA1G
  • Clonality:
    Monoclonal
  • Clone:
    S178A-9
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    12291
  • Antigen synonyms:
    Cav3.1d|[a]1G|mKIAA1123|a1G|alpha-1G
  • Amino acid number:
    2052 to 2172
  • Storage buffer:
    PBS, pH 7,4 (50% glycerol)
  • Storage temperature:
    Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant fusion protien corresponding to amino acids 2052-2172 at C-terminus of mouse Cacna1g.
  • Purification:
    Protein G Purified
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against recombinant of Cacna1g.

Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000

Type: Primary
Antigen: CACNA1G
Clonality: Monoclonal
Clone: S178A-9
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: