Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Solute carrier family 17, member A6
- Antigen symbol:SLC17A6
- Clonality:Monoclonal
- Clone:S29-29
- Conjugation:RPE (R-Phycoerythrin)
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Format:Liquid
- Gene ID:84487
- Antigen synonyms:Vglut2|Dnpi
- Amino acid number:501 to 582
- Storage buffer:PBS, pH 7,4 (50% glycerol)
- Storage temperature:Store at 4 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant fusion protien corresponding to amino acids 501-582 at C-terminus of rat Slc17a6.
- Purification:Protein G Purified
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against recombinant of Slc17a6.
Recommended Dilutions: Immunohistochemistry Western Blot: 1:1000
Type: Primary
Antigen: SLC17A6
Clonality: Monoclonal
Clone: S29-29
Conjugation: RPE (R-Phycoerythrin)
Epitope: C-Terminal
Host: Mouse
Isotype: IgG1
Reactivity: