Anti-ICAM1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB25041
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Intercellular Adhesion molecule 1
- Antigen symbol:ICAM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:3383
- Antigen synonyms:ICAM-1|major group rhinovirus receptor|human rhinovirus receptor|BB2|cell surface glycoprotein P3.58|intercellular adhesion molecule 1|P3.58
- Modification:phospho-Tyr512
- Storage buffer:PBS, pH 7,2 (0,05% sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Synthetic phosphopeptide corresponding to residues surrounding Y512 of ICAM1.
- Purification:Antigen affinity purification
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic phosphopeptide of ICAM1.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human