Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD38 molecule
- Antigen symbol:CD38
- Clonality:Monoclonal
- Clone:6E12D
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Form:Ascites
- Gene ID:952
- Antigen synonyms:NAD+ nucleosidase|ADP-ribosyl cyclase 1|T10|cADPr hydrolase 1|ADP-ribosyl cyclase/cyclic ADP-ribose hydrolase|cyclic ADP-ribose hydrolase 1|CD38 antigen p45
- Storage buffer:0,03% sodium azide
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to human CD38.
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Mouse monoclonal antibody raised against partial recombinant CD38.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:500-1:2000; Immunohistochemistry: 1:200-1:1000
Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 6E12D
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human