Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Cadherin 2
- Antigen symbol:CDH2
- Clonality:Monoclonal
- Clone:3G3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Format:Liquid
- Gene ID:1000
- Antigen synonyms:CDHN|Neural-Cadherin|CDw325|N-Cadherin|CDH2|NCAD
- Amino acid number:807 to 906
- Storage buffer:1x PBS, pH 7,4
- Sequence:RRMDERPIHAEPQYPVRSAAPHPGDIGDFINEGLKAADNDPTAPPYDSLLVFDYEGSGSTAGSLSSLNSSSSGGEQDYDYLNDWGPRFKKLADMYGGGDD
- Storage temperature:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:CDH2 (NP_001783, 807 a.a. ~ 906 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant CDH2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: CDH2
Clonality: Monoclonal
Clone: 3G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human