Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:CD59 Molecule, Complement Regulatory Protein
- Antigen symbol:CD59
- Clonality:Monoclonal
- Clone:MEM-43
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:966
- Antigen synonyms:MIRL|HRF-20|MEM43|1F5|MACIF|MIN3|EJ16|MSK21|EL32|HRF20|p18-20|MIC11|MIN1|16.3A5|MIN2|MAC-IP|G344|EJ30
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified CD59 from thymocytes and T lymphocytes.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against native CD59.
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: MEM-43
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human