Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-HLAA Mouse Monoclonal Antibody [clone: MEM-81]
Anti-HLAA Mouse Monoclonal Antibody [clone: MEM-81]
Catalog # ABNOMAB0904
Supplier:  Abnova
Anti-HLAA Mouse Monoclonal Antibody [clone: MEM-81]
Catalog # ABNOMAB0904
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    HLAA
  • Clonality:
    Monoclonal
  • Clone:
    MEM-81
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    3105|3106|3107
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified HLA-Class I from leukocytes of a patient suffering from a LGL-type leukemia.
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native HLA-Class I.

Type: Primary
Antigen: HLAA
Clonality: Monoclonal
Clone: MEM-81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human