Order Entry
Northern Ireland
ContactUsLinkComponent
IgG1 Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))
IgG1 Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOMAB10143
Supplier:  Abnova
IgG1 Isotype Control [clone: MOPC-21] (FITC (Fluorescein Isothiocyanate))
Catalog # ABNOMAB10143
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    MOPC-21
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    PBS, pH 7,4 (15 mM sodium azide)
  • Storage temperature:
    Store at 4 °C. Do not freeze.
  • Purification:
    Protein G purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody was chosen by screening on variety of resting, activated, live and fixed rat and human tissues.

Type: Control
Antigen: IgG1
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host:
Isotype: IgG1
Reactivity: