Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Antibody type:Primary
- Antigen name:Membrane-spanning 4-Domains, Subfamily A, Member 1
- Antigen symbol:MS4A1
- Clonality:Monoclonal
- Clone:2H7
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Primates,Human
- Cross adsorption:No
- Format:Liquid
- Gene ID:931
- Antigen synonyms:CVID5|Bp35|S7|LEU-16|MS4A2|CD20|B1
- Storage buffer:PBS (0,2% BSA, 15 mM sodium azide)
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Human tonsillar B cells.
- Purification:Size-exclusion chromatography purification
- Size:100 Reactions
- Pk:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against MS4A1.
Recommended Dilutions: Flow Cytometry: 0.6 ?g/ml
Type: Primary
Antigen: MS4A1
Clonality: Monoclonal
Clone: 2H7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Non-human Primates