Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 28-14-8]
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 28-14-8]
Catalog # ABNOMAB3258
Supplier:  Abnova
Anti-H2-D2 Mouse Monoclonal Antibody [clone: 28-14-8]
Catalog # ABNOMAB3258
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    H2-D2
  • Clonality:
    Monoclonal
  • Clone:
    28-14-8
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Mouse
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    100 mM BBS, pH 8,2
  • Storage temperature:
    Store at 4 °C.
  • Immunogen:
    Native from C3H.SW mouse skin graft and splenocytes.
  • Pk:
    500 µG

Specifications

About this item

Mouse monoclonal antibody raised against H2-D2.

Recommended Dilutions: Flow Cytometry: 1 ?g/10^6 cells

Type: Primary
Antigen: H2-D2
Clonality: Monoclonal
Clone: 28-14-8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse