Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81]
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81]
Catalog # ABNOMAB2392
Supplier:  Abnova
Anti-TRA-1-81 Mouse Monoclonal Antibody [clone: TRA-1-81]
Catalog # ABNOMAB2392
Supplier:  Abnova

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    TRA-1-81
  • Clonality:
    Monoclonal
  • Clone:
    TRA-1-81
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Storage buffer:
    buffer containing 0,1% sodium azide
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C or –80 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Native purified human embryonal carcinoma cell line 2102Ep.
  • Size:
    100 μl
  • Pk:
    100 µg

Specifications

About this item

Mouse monoclonal antibody raised against native TRA-1-81.

Recommended Dilutions: Flow Cytometry: 1:50-1:200; Immunocytochemistry: 1:50-1:200

Type: Primary
Antigen: TRA-1-81
Clonality: Monoclonal
Clone: TRA-1-81
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human